ContactUsLinkComponent
Recombinant protein, blk( aah07371, 1 a.a. - 90 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000640-Q01
Supplier:  Abnova
Recombinant protein, blk( aah07371, 1 a.a. - 90 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000640-Q01
Supplier:  Abnova