ContactUsLinkComponent
Recombinant protein, bbs1( np_078925, 187 a.a. - 295 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000582-Q01
Supplier:  Abnova
Recombinant protein, bbs1( np_078925, 187 a.a. - 295 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000582-Q01
Supplier:  Abnova