ContactUsLinkComponent
Recombinant protein, atr( np_001175, 2545 a.a. - 2644 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000545-Q01
Supplier:  Abnova
Recombinant protein, atr( np_001175, 2545 a.a. - 2644 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000545-Q01
Supplier:  Abnova