ContactUsLinkComponent
Recombinant protein, hmgn4( aah01282, 1 a.a. - 91 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010473-P01
Supplier:  Abnova
Recombinant protein, hmgn4( aah01282, 1 a.a. - 91 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010473-P01
Supplier:  Abnova