ContactUsLinkComponent
Recombinant protein, tagln( aah04927, 1 a.a. - 202 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006876-P01
Supplier:  Abnova
Recombinant protein, tagln( aah04927, 1 a.a. - 202 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006876-P01
Supplier:  Abnova