ContactUsLinkComponent
Recombinant protein, znf226( np_001027544.1, 1 a.a. - 803 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
  ABNOH00007769-P01
 :  Abnova
Recombinant protein, znf226( np_001027544.1, 1 a.a. - 803 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
  ABNOH00007769-P01
 :  Abnova
Frequently Bought Together