ContactUsLinkComponent
Recombinant protein, znf133( aah01887, 1 a.a. - 654 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007692-P01
Supplier:  Abnova
Recombinant protein, znf133( aah01887, 1 a.a. - 654 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007692-P01
Supplier:  Abnova