ContactUsLinkComponent
Recombinant protein, pin1( np_006212.1, 1 a.a. - 163 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00005300-P01
Supplier:  Abnova
Recombinant protein, pin1( np_006212.1, 1 a.a. - 163 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00005300-P01
Supplier:  Abnova