ContactUsLinkComponent
Recombinant protein, dnm2( np_001005360.1, 1 a.a. - 870 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001785-P01
Supplier:  Abnova
Recombinant protein, dnm2( np_001005360.1, 1 a.a. - 870 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001785-P01
Supplier:  Abnova