ContactUsLinkComponent
Recombinant protein, gtf2h1( aah04452, 1 a.a. - 549 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00002965-P01
Supplier:  Abnova
Recombinant protein, gtf2h1( aah04452, 1 a.a. - 549 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00002965-P01
Supplier:  Abnova