ContactUsLinkComponent
Recombinant protein, nt5e( aah15940.1, 28 a.a. - 265 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00004907-P01
Supplier:  Abnova
Recombinant protein, nt5e( aah15940.1, 28 a.a. - 265 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00004907-P01
Supplier:  Abnova