ContactUsLinkComponent
Recombinant protein, gck( aah01890, 1 a.a. - 466 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002645-P01
Supplier:  Abnova
Recombinant protein, gck( aah01890, 1 a.a. - 466 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002645-P01
Supplier:  Abnova