ContactUsLinkComponent
Recombinant protein, gclc( np_001489.1, 1 a.a. - 637 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002729-P01
Supplier:  Abnova
Recombinant protein, gclc( np_001489.1, 1 a.a. - 637 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002729-P01
Supplier:  Abnova