SUPORLIFE 200 DCF filter with a double layer polyethersulfone membrane and an absolute removal rating of 0.2um. Supplied with 1/4 inch MNPT inlet/outlet connections with vent
Catalog # PALBCHS92DPDDL
Supplier: Palbam
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.