ContactUsLinkComponent
STIRRED BATH/CIRCULATOR, OPTIMA TX150 PROGRAMMABLE IMMERSION THERMOSTAT + ST5 STAINLESS STEEL TANK, 5L CAPACITY. TEMP RANGE AMBIENT +15 TO 150C. INCL. PUMP. 1 * 1 items
Catalog # 461-0710
Supplier:  GRANT INSTRUMENTS
STIRRED BATH/CIRCULATOR, OPTIMA TX150 PROGRAMMABLE IMMERSION THERMOSTAT + ST5 STAINLESS STEEL TANK, 5L CAPACITY. TEMP RANGE AMBIENT +15 TO 150C. INCL. PUMP. 1 * 1 items
Catalog # 461-0710
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together