ContactUsLinkComponent
Stirred bath/circulator, optima tx150 programmable immersion thermostat + p5 plastic tank, 5l capacity. temp range amb. +15 to 99c. incl. pump. 1 * 1 items
Catalog # GRANTX150-P5EU
Supplier:  GRANT INSTRUMENTS
Stirred bath/circulator, optima tx150 programmable immersion thermostat + p5 plastic tank, 5l capacity. temp range amb. +15 to 99c. incl. pump. 1 * 1 items
Catalog # GRANTX150-P5EU
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together