Thermo Scientific F1 ClipTip 5ul, single channel, fixed volume, fits with Thermo Scientific 20ul ClipTip pipette tip, features new l ow force attachment and ejection mechanism and secured tip attachment system 1 * 1 items
Catalog # 613-2185
Supplier: Thermo Fisher Scientific
Frequently Bought Together