ContactUsLinkComponent
Flask cell culture polystyrene working volume 60ml cell growth area 75cm rectangular canted neck with orange plug seal cap graduated TC-treated sterile 1 * 5 items
Catalog # CORN430720.IGC/5
Supplier:  Corning
Flask cell culture polystyrene working volume 60ml cell growth area 75cm rectangular canted neck with orange plug seal cap graduated TC-treated sterile 1 * 5 items
Catalog # CORN430720.IGC/5
Supplier:  Corning
Frequently Bought Together