Fixed Angle Rotor for 30 x 1.5 / 2.0ml Microtubes, Max Speed:15,000rpm /24,400g, With Aerosol-Tight lid, Rotor Angle: 45, Autoclavable to 136 C. 1 * 1 items
Catalog # HETT4489-A
Supplier: Hettich
Fixed Angle Rotor for 30 x 1.5 / 2.0ml Microtubes, Max Speed:15,000rpm /24,400g, With Aerosol-Tight lid, Rotor Angle: 45, Autoclavable to 136 C. 1 * 1 items
Catalog # HETT4489-A
Supplier: Hettich
Frequently Bought Together
Pipette controller, PIPETBOY acu 2
Supplier: Integra
VWR®, Stem Thermometer, Calibrated, Electronic, Blunt Tip, Stainless Steel, 120 mm Probe
Supplier: VWR Collection
Finnpipette™ F1 Single Channel Pipettes, Mechanical, Fixed/Variable Volume
Supplier: Thermo Fisher Scientific
Accessories for marco
Supplier: Brand
Single-channel pipettes, mechanical, fixed / variable volume, Finnpipette™ F2
Supplier: Thermo Fisher Scientific
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Microlab 600 Bubble Free Prime (BFP) syringes
Supplier: HAMILTON BONADUZ
VWR® Traceable®, Waterproof Thermometer
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWR®, Environmentally Friendly Thermometers, Easy Read
Supplier: VWR Collection
VWR® Probeless Electronic Thermometers
Supplier: VWR Collection
VWR® Traceable® Alarm Thermometer with Timer
Supplier: VWR Collection
VWR® Calibratable Ultra-Accurate Thermometers
Supplier: VWR Collection
VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 120 mm Probe
Supplier: VWR Collection
Recirculating coolers, F series
Supplier: JULABO GmbH
Accessories for aspirators, GRANT INSTRUMENTS
Supplier: GRANT INSTRUMENTS
Accessories
Supplier: MIELE
PRINTER RIBBON CARTRIDGE f. CBM-910II (Citizen) 1 * 1 items
Supplier: SHP Steriltechnik