Orders LinkContactUsLinkComponent
Examination light
Examination light
Examination light, HALUX LED 20 P LX, UK-plug
Catalog # 139-0184
Supplier:   WALDMANN
Examination light
Catalog # 139-0184
Supplier:  WALDMANN
Examination light
Examination light
Examination light, HALUX LED 20 P LX, UK-plug
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Powerful down to the smallest details, it does not diffuse any infrared radiation - no heat. A twin-wall body including fan optimizes cooling of luminaire head. A simple control unit is integrated on luminaire head. Friction system for a risk-free and precise light positioning.
  • Standard adapter for different fixture types
  • Electronic transformer
  • Robust and ergonomic design
  • Long LED service life: Reduced maintenance costs
  • Easy cable replacement: Device connector on transformer housing
IEC 60601-2-41 standard examination light
Specifications
  • Description:
    Examination light, HALUX LED 20 P LX, UK-plug