ContactUsLinkComponent
Hydrolysis Unit B-411, 230 V, VITON 4-place unit equipped with specially robust rubber couplings made from Viton. Up to 4 samples ca n be hydrolyzed simultaneously. Scope of deliver 1 * 1 items
Catalog # BUCH037461
Supplier:  BUCHI
Hydrolysis Unit B-411, 230 V, VITON 4-place unit equipped with specially robust rubber couplings made from Viton. Up to 4 samples ca n be hydrolyzed simultaneously. Scope of deliver 1 * 1 items
Catalog # BUCH037461
Supplier:  BUCHI
Frequently Bought Together