Inlet base seals
Gold-plated inlet base seals, 0,8 mm ID, for use with single column installation
Catalog # 555-9038
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.
Specifications
- Description:Gold-plated inlet base seals, 0,8 mm ID, for use with single column installation
- Seq:01