Orders LinkContactUsLinkComponent
Inlet base seals
Inlet base seals
Gold-plated cross inlet base seals, 0,8 mm ID
  555-9223
Inlet base seals
  555-9223
Inlet base seals
Inlet base seals
Gold-plated cross inlet base seals, 0,8 mm ID
£333.00
1 * 10 items
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.
  • Description:
    Gold-plated cross inlet base seals, 0,8 mm ID
  • Seq:
    06