ContactUsLinkComponent
safety cabinet with wing door. outer carcass: 595 x 745 x 2080 (wxdxh) mm, inner carcass: 499 x 595 x 1830 (wxdxh) mm, wing door hun g right outer carcass, sheet steel, powder-coated in ral 7035 light grey, wing doors in ral 1018 zinc yellow, inner c 1 * 1 items
Catalog # DUEH29-200670-081
Supplier:  DUPERTHAL
safety cabinet with wing door. outer carcass: 595 x 745 x 2080 (wxdxh) mm, inner carcass: 499 x 595 x 1830 (wxdxh) mm, wing door hun g right outer carcass, sheet steel, powder-coated in ral 7035 light grey, wing doors in ral 1018 zinc yellow, inner c 1 * 1 items
Catalog # DUEH29-200670-081
Supplier:  DUPERTHAL