ContactUsLinkComponent
Developing chamber is extremely useful for TLC. Consists of a sturdy, molded clear glass block with flat surface walls and ground lip of uniform flatness. Raised ridge midway on inside ends and bottom of chamber. Holds up to six 20x20cm plates. Chamb 1 * 1 items
Catalog # VWRMNP80-10
Supplier:  VWR Products
Developing chamber is extremely useful for TLC. Consists of a sturdy, molded clear glass block with flat surface walls and ground lip of uniform flatness. Raised ridge midway on inside ends and bottom of chamber. Holds up to six 20x20cm plates. Chamb 1 * 1 items
Catalog # VWRMNP80-10
Supplier:  VWR Products
Frequently Bought Together