ContactUsLinkComponent
n6-(benzyloxycarbonyl)-l-lysine is a protected amino acid. cas number: 1155-64-2 synonym: n-z-l-lysine, n6-carbobenzyloxy-l-lysine linear formula: 6h5ch2oocnh(ch2)4ch(nh2)cooh molecular weight: 280.32 1 * 25 g
Catalog # GENO786-550
Supplier:  G-Biosciences
n6-(benzyloxycarbonyl)-l-lysine is a protected amino acid. cas number: 1155-64-2 synonym: n-z-l-lysine, n6-carbobenzyloxy-l-lysine linear formula: 6h5ch2oocnh(ch2)4ch(nh2)cooh molecular weight: 280.32 1 * 25 g
Catalog # GENO786-550
Supplier:  G-Biosciences