ContactUsLinkComponent
Stainless Steel Bottle. Volume: 110mL. Constructed of T316L stainless steel. With 5.1cm tri-clamp and 12 Ra electropolished finish. Standard gasket material is platinum-cured silicone. Designed for pilot/stability testing of small quantities of produ 1 * 1 items
Catalog # EAGLSTBEP110316L
Stainless Steel Bottle. Volume: 110mL. Constructed of T316L stainless steel. With 5.1cm tri-clamp and 12 Ra electropolished finish. Standard gasket material is platinum-cured silicone. Designed for pilot/stability testing of small quantities of produ 1 * 1 items
Catalog # EAGLSTBEP110316L
Frequently Bought Together