Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

Buffer, TE Solution (TRIS-EDTA buffer) pH 8,0, Low EDTA for Molecular Biology

TE buffer solution (TRIS-EDTA buffer) pH 8,0, Glass bottle, 1 L
  A8569.1000
Buffer, TE Solution (TRIS-EDTA buffer) pH 8,0, Low EDTA for Molecular Biology
  A8569.1000
 :  
Buffer, TE Solution (TRIS-EDTA buffer) pH 8,0, Low EDTA for Molecular Biology

TE buffer solution (TRIS-EDTA buffer) pH 8,0, Glass bottle, 1 L

 :  
 
£54.70
   
1 * 1 L
 
  • DNases/RNases/Proteases: not detectable
  • pH (20°C; adjusted with HCl): 8,0 ± 0,1

Composition: EDTA · Na2: 0,1 mM and Tris: 10 mM.

 

 

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TE buffer solution (TRIS-EDTA buffer) pH 8,0