RAZOR BLADE DOUBLE EDGE 0.23MM All blades are industrial grade. Single-edge blades are 0.23mm (0.009 ) thick and feature an extra-keen edge. Blades are available with steel or aluminum spines. Double-edge blades are 0.1mm(0.004 ) thick and are oiled 1 * 250 items
Catalog # AMSR95-0043
Supplier: AMERICAN SAFETY RAZOR
Frequently Bought Together