ContactUsLinkComponent
Eppendorf 5430 Ambient Microcentrifuge - Dial/Knob Version Supplied without rotor Max. Capacity: 48 x 1.5 /2.0 ml Max Speed/RCF: 17,500 rpm/30,130 g Brushless Induction Motor Microprocessor Control with Large, Backlit Digital Display Timer: 30s to 9 1 * 1 items
Catalog # EPPE5427000.666
Supplier:  EPPENDORF
Eppendorf 5430 Ambient Microcentrifuge - Dial/Knob Version Supplied without rotor Max. Capacity: 48 x 1.5 /2.0 ml Max Speed/RCF: 17,500 rpm/30,130 g Brushless Induction Motor Microprocessor Control with Large, Backlit Digital Display Timer: 30s to 9 1 * 1 items
Catalog # EPPE5427000.666
Supplier:  EPPENDORF
Frequently Bought Together