ContactUsLinkComponent
Eppendorf 5430 Ambient Microcentrifuge - Dial/Knob Version Supplied with Rotor for 30 x 1.5/2.0 ml Microtubes, PTFE coating and and QuickLock Aerosol-tight Lid Max. Capacity: 48 x 1.5 /2.0 ml Max Speed/RCF: 17,500 rpm/30,130 g Brushless Induction Mot 1 * 1 items
Catalog # 521-1563
Supplier:  EPPENDORF
Eppendorf 5430 Ambient Microcentrifuge - Dial/Knob Version Supplied with Rotor for 30 x 1.5/2.0 ml Microtubes, PTFE coating and and QuickLock Aerosol-tight Lid Max. Capacity: 48 x 1.5 /2.0 ml Max Speed/RCF: 17,500 rpm/30,130 g Brushless Induction Mot 1 * 1 items
Catalog # 521-1563
Supplier:  EPPENDORF
Frequently Bought Together