ULTRA STREPTAVIDIN-HRP 1 * 1,5 mL
Catalog # PIERN504
Supplier: Thermo Fisher Scientific
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Ethylene glycol ≥99% (by GC), pure
Supplier: PanReac AppliChem
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 98%, pure
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
1-Chloro-2,4-dinitrobenzene 99%
Supplier: Thermo Fisher Scientific