pH7 Buffer Solution 500ml ##Bottle,Yellow. PHM117 788-14 1 * 1 items
Catalog # SENK788-14
Supplier: SENTEK
pH7 Buffer Solution 500ml ##Bottle,Yellow. PHM117 788-14 1 * 1 items
Catalog # SENK788-14
Supplier: SENTEK
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck