Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Bismuth(III) carbonate basic

Bismuth(III) carbonate basic

Glass bottle for solids, 250 g
Catalog # A12483.30
Bismuth(III) carbonate basic
Catalog # A12483.30
CAS Number:  5892-10-4
Bismuth(III) carbonate basic
Bismuth(III) carbonate basic

Glass bottle for solids, 250 g

CAS Number:  5892-10-4
Price
£90.20
Log in for availability
1 * 250 g
Product Details & Documents

CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 Formula Weight: 509.97 Merck Reference: 14,1282 MDL No.: MFCD00010877

Synonyms:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS Number:5892-10-4
  • Formula:(BiO)₂CO₃
  • MDL Number:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Merck Index:14,1282
  • Density:6,86
  • Storage Temperature:Ambient
  • EINECS:227-567-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.