Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 10 mm Ø tube, 20
Catalog # JENC357-128
Supplier: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Catalog # JENC357-128
Supplier: JENCONS SCIENTIFIC
Jump to:
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifications
- No. of Holes:20
- Description:Interchangeable block
- Seq:07
- For:10 mm Ø tube