Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Trays, large deep

Trays, large deep

White, 65 l, 630×440×340 mm
Catalog # DELT19604
Supplier:   DELTALAB
Trays, large deep
Catalog # DELT19604
Supplier:  DELTALAB
Trays, large deep

White, 65 l, 630×440×340 mm

Price
£111.00
Log in for availability
1 * 1 items
Product Details & Documents

Trays made of natural HDPE.

  • Stackable
  • Good temperature resistance
  • Good chemical resistance

With ergonomically designed handles.

Specifications
  • Capacity:
    65 l
  • Colour:
    White
  • Seq:
    02
  • W×D×H:
    630×440×340 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.