Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CAP BAKELITE TO FIT WM BOTTLE 30ML 1 * 10 items
Catalog # MURRG8080710
Supplier:
MURRAY S
Expand image
CAP BAKELITE TO FIT WM BOTTLE 30ML 1 * 10 items
Catalog # MURRG8080710
Supplier:
MURRAY S
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Funnels with Short Stem
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Control seal, close-it tape
Supplier: BURKLE