ContactUsLinkComponent
Areometer, DIN12791, L50 series, measuring range:0.70-0.75:0.0005g/cm , without thermometer, reference temperature:20C, length:335m m, suitable for government verification. 1 * 1 items
Catalog # AMARH806204
Supplier:  Amarell
Areometer, DIN12791, L50 series, measuring range:0.70-0.75:0.0005g/cm , without thermometer, reference temperature:20C, length:335m m, suitable for government verification. 1 * 1 items
Catalog # AMARH806204
Supplier:  Amarell
Frequently Bought Together