ContactUsLinkComponent
Mouse Developmental Tissue qPCR Array containing first strand DNA from 48 tissues (2 panels) at five stages (3 embryo, 1 post natal and 1 adult) 1 * 1 items
Catalog # ORIGMDRT101
Supplier:  OriGene
Mouse Developmental Tissue qPCR Array containing first strand DNA from 48 tissues (2 panels) at five stages (3 embryo, 1 post natal and 1 adult) 1 * 1 items
Catalog # ORIGMDRT101
Supplier:  OriGene
Frequently Bought Together