CHLOROPHENOL COCKTAIL SOLUTION 1 * 1,2 mL
PROOCIL-EM-1727-A
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
: VWR Chemicals
Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
: Thermo Fisher Scientific
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
: Thermo Fisher Scientific