Orders LinkContactUsLinkComponent
RNaseOUT™ RNAse removal solution
RNaseOut™, spray bottle, 250 mL
  786-70
RNaseOUT™ RNAse removal solution
  786-70
 :  
RNaseOUT™ RNAse removal solution
RNaseOUT™ RNAse removal solution
RNaseOut™, spray bottle, 250 mL
 :  
£47.80
1 * 250 mL
RNaseOUT™ is uniquely formulated to destroy and remove RNases on contact.
  • Simply spray and rinse
  • RNaseOUT™ is non-toxic and residue free
  • RNaseOUT™ allows common equipment to be safely and confidently used for all RNA work
  • Pk:
    250 mL
  • Description:
    RNaseOut™, spray bottle