n-Hexane ≥95%, TECHNICAL
Metal drum, 204 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–94,3 °C
- Density:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
- REACH:01-2119480412-44
Specifications
Specification Test Results
- Assay:Min. 95 %
- Boiling point:68 - 70 °C
- Benzene:Max. 100 ppm
- Evaporation residue:Max. 20 ppm
Specifications
- Pack type:Metal drum
- Pk:204 L



