ContactUsLinkComponent
Fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem and CH-plugChamberdimension (xL) 310 x 700 mm Volume: 55 liters 1 * 1 items
Catalog # 481-2030
Supplier:  TUTTNAUER
Fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem and CH-plugChamberdimension (xL) 310 x 700 mm Volume: 55 liters 1 * 1 items
Catalog # 481-2030
Supplier:  TUTTNAUER
Frequently Bought Together