FUSE TEMPERATURE 110C (-7K) 1 * 1 items
Catalog # BIND5006-0043
Supplier: Binder
Frequently Bought Together
Accessories for accu-jet® pro
Supplier: Brand
Filter protection 5 ml 1 * 50 items
Supplier: EPPENDORF
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Accessories for Multipette® M4
Supplier: EPPENDORF
Accessories for Research® Plus pipettes
Supplier: EPPENDORF
Sacrifical body 1 * 1 items
Supplier: VWR Collection
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for Titrette®
Supplier: Brand
Accessories for HandyStep® electronic
Supplier: Brand
Filter,membrane for research pipette 1 * 50 items
Supplier: EPPENDORF
Accessories for CO2 Incubators, Heracell VIOS
Supplier: Thermo Scientific
Door Gasket For VT6025 Oven 1 * 1 items
Supplier: Thermo Scientific
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
Accessories for water purification millipore
Supplier: Merck
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for dispersing elements for homogenisers
Supplier: IKA
ORP combination electrodes, IntelliCAL™ MTC101 / MTC301
Supplier: Hach
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach