ContactUsLinkComponent
Thermometer, general purpose, simple type, red special liquid filled, measuring range:-10:110/1C, length:300mm, diameter:6-7mm, 76m m immersion, solid stem, white backed, with ring at top, durable pigment. 1 * 1 items
Catalog # AMARG11672
Supplier:  Amarell
Thermometer, general purpose, simple type, red special liquid filled, measuring range:-10:110/1C, length:300mm, diameter:6-7mm, 76m m immersion, solid stem, white backed, with ring at top, durable pigment. 1 * 1 items
Catalog # AMARG11672
Supplier:  Amarell
Frequently Bought Together