Multiple adapters
10/19 NS, 10/19 NS
Catalog # SMIT8SMAP/00
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
Borosilicate glass
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Multiple neck, two parallel necks.
Specifications
- Cone size:10/19 NS
- Socket size:10/19 NS
- Seq:01