Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Pestles, serrated

Pestles, serrated

230 mm, 2 ml mortars, 6,5 mm
Catalog # KART6302
Supplier:   KARTELL
Pestles, serrated
Catalog # KART6302
Supplier:  KARTELL
Pestles, serrated
Pestles, serrated

230 mm, 2 ml mortars, 6,5 mm

Price
£65.10
Log in for availability
1 * 1 items
Product Details & Documents

Pestles used in the combination with borosilicate mortar for sample crushing. Ideal for tough materials such as tumours.

  • Head made of PTFE
  • Shaft made of stainless steel
Specifications
  • Ø:
    6,5 mm
  • For:
    2 ml mortars
  • Length:
    230 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.