Orders LinkContactUsLinkComponent
Büchner funnels
Büchner funnels
97 mm, 90 mm
Catalog # SMIT237/4
Supplier:   SMITH SCIENTIFIC
Büchner funnels
Catalog # SMIT237/4
Supplier:  SMITH SCIENTIFIC
Büchner funnels
Büchner funnels
97 mm, 90 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
In white glazed porcelain.
For paper or membrane filtration discs.
Specifications
  • Plate Ø:
    97 mm
  • For filter Ø:
    90 mm