Büchner funnels
62 mm, 55 mm
Catalog # SMIT237/2
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
In white glazed porcelain.
For paper or membrane filtration discs.
Specifications
- Plate Ø:62 mm
- For filter Ø:55 mm
In white glazed porcelain.
For paper or membrane filtration discs.