Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

Boc-ON ≥98%

25 g
Catalog # A18906.14
Irritant
Boc-ON ≥98%
Catalog # A18906.14
CAS Number:  58632-95-4
Boc-ON ≥98%
Boc-ON ≥98%

25 g

CAS Number:  58632-95-4
Price
£89.40
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 58632-95-4 Molecular Formula: C13H14N2O3 Formula Weight: 246.27 Storage Temperature: 2°C to 8°C Melting Point: 86-89° Beilstein Reference: 2217296 UN No.: UN3439 MDL No.: MFCD00001863

Synonyms:  2-(tert-BOC-Oxyimino)-2-phenylacetonitrile, N-((tert-Butoxycarbonyl)oxy)benzimidoyl cyanide, 2-(Boc-oxyimino)-2-phenylacetonitrile, 2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile, [(cyano-phenyl-methylidene)amino] tert-butyl carbonate
  • CAS Number:58632-95-4
  • Formula:C₁₃H₁₄N₂O₃
  • MDL Number:MFCD00001863
  • Molecular Weight:246.27 g/mol
  • Melting Pt:86…89 °C
  • EINECS:261-370-9
  • ADR:6.1,III
Specifications

Specifications

  • Pk:
    25 g

Specifications